Learn More
N-Phthaloylglycine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | N-Phthaloylglycine |
| CAS | 4702-13-0 |
| Molecular Formula | C10H7NO4 |
| Quantity | 25 g |
| Synonym | 1,3-Dihydro-1,3-dioxo-2H-isoindole-2-acetic Acid, 1,3-Dioxo-2-isoindolineacetic Acid, (1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetic Acid, (1,3-Dioxo-1,3-dihydroisoindol-2-yl)acetic Acid, 1,3-Dihydro-1,3-dioxo-2H-isoindole-2-acetic Acid, 1,3-Dioxo-2-isoindolineacetic Acid, 2-(1,3-Dioxoisoindolin-2-yl)acetic Acid, 2-Phthalimidoacetic Acid, N-(2-Hydroxy-2-oxoethyl)phthalimide, N-(Carboxymethyl)phthalimide, NSC 10771, NSC 29044, Phthalimidoacetic Acid, Phthaloglycine, Phthaloylglycine |
| InChI Formula | InChI=1S/C10H7NO4/c12-8(13)5-11-9(14)6-3-1-2-4-7(6)10(11)15/h1-4H,5H2,(H,12,13) |
| SMILES | OC(=O)CN1C(=O)c2ccccc2C1=O |
| IUPAC Name | 2-(1,3-dioxoisoindol-2-yl)acetic acid |
| Molecular Weight (g/mol) | 205.17 |
| Formula Weight | 205.0375 |