Learn More
N-Phthaloylglycine, 98+%
Product Code. 11458456
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
Product Code. 11458456
Brand: Thermo Scientific Alfa Aesar A15920.30
Request Bulk or Custom Format
CAS: 4702-13-0 | C10H7NO4 | 205.169 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4702-13-0
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.169
MDL Number MFCD00005900
InChI Key WQINSVOOIJDOLJ-UHFFFAOYSA-N
Synonym n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid
PubChem CID 20825
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O
Specifications
Melting Point 194°C to 198°C
Beilstein 184174
Quantity 250 g
Formula Weight 205.17
Percent Purity ≥98%
Chemical Name or Material N-Phthaloylglycine