Learn More
N-Phenyliminodiacetic acid, 97%
Product Code. 11388165
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11388165
Brand: Thermo Scientific Alfa Aesar L08437.06
Request Bulk or Custom Format
CAS: 1137-73-1 | C10H11NO4 | 209.201 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1137-73-1
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.201
MDL Number MFCD00043570
InChI Key GQBWTAGIANQVGB-UHFFFAOYSA-N
Synonym n-phenyliminodiacetic acid, glycine, n-carboxymethyl-n-phenyl, acetic acid,1-phenyliminodi, 2,2'-phenylazanediyl diacetic acid, 2,2'-phenylimino diacetic acid, n-carboxymethyl-n-phenylglycine, n,n-anilinediacetic acid, n-phenyliminodiacetic acid anhydride, nn-di carboxymethyl aniline, glycine,n-carboxymethyl-n-phenyl
PubChem CID 136915
IUPAC Name 2-[N-(carboxymethyl)anilino]acetic acid
SMILES C1=CC=C(C=C1)N(CC(=O)O)CC(=O)O
Specifications
Melting Point ∽140°C (decomposition)
Quantity 5 g
Beilstein 2808435
Formula Weight 209.2
Percent Purity 97%
Chemical Name or Material N-Phenyliminodiacetic acid