Learn More
N-Phenylglycinonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | N-Phenylglycinonitrile |
| CAS | 3009-97-0 |
| Molecular Formula | C8H8N2 |
| Quantity | 250 g |
| Synonym | 2-(Phenylamino)acetonitrile, (Phenylamino)acetonitrile, 1-Anilinoacetonitrile, Anilinoacetonitrile, N-(Cyanomethyl)aniline, N-(Phenylamino)acetonitrile, Phenylglycine Nitrile, NSC 7611 |
| InChI Formula | InChI=1S/C8H8N2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,10H,7H2 |
| SMILES | N#CCNc1ccccc1 |
| IUPAC Name | 2-anilinoacetonitrile |
| Molecular Weight (g/mol) | 132.16 |
| Formula Weight | 132.0687 |