missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Phenylglycine, 97%
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Beskrivning
N-Phenylglycine coupled with Glycidyl methacrylate to create a surface active comonomer that promotes adhesive bonding of materials to hard tooth tissues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Phenylglycine coupled with Glycidyl methacrylate to create a surface active comonomer that promotes adhesive bonding of materials to hard tooth tissues.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
N-Phenylglycine coupled with Glycidyl methacrylate to create a surface active comonomer that promotes adhesive bonding of materials to hard tooth tissues.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifikationer
Specifikationer
| Melting Point | 117°C to 119°C |
| Beilstein | 509838 |
| Merck Index | 14,7292 |
| Solubility Information | Soluble in water. |
| Quantity | 50 g |
| Formula Weight | 151.17 |
| Percent Purity | 97% |
| Chemical Name or Material | N-Phenylglycine |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur