missing translation for 'onlineSavingsMsg'
Learn More

n-Pentylbenzene, 96%

Product Code. 11402378
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11402378

Brand: Thermo Scientific Alfa Aesar A15734.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 538-68-1 | C11H16 | 148.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-68-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00009502
InChI Key PWATWSYOIIXYMA-UHFFFAOYSA-N
Synonym 1-phenylpentane, n-pentylbenzene, amylbenzene, n-amylbenzene, benzene, pentyl, 1-pentylbenzene, 1-phenyl-n-pentane, pentane, 1-phenyl, pentyl-benzene, pentane, phenyl
PubChem CID 10864
IUPAC Name pentylbenzene
SMILES CCCCCC1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.863
Boiling Point 204°C to 205°C
Flash Point 65°C (149°F)
Refractive Index 1.488
Quantity 100 g
UN Number UN3082
Beilstein 1904359
Merck Index 14,600
Formula Weight 148.25
Percent Purity 96%
Chemical Name or Material n-Pentylbenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.