missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentyl Alcohol-d12, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Pentanol-D12 |
| CAS | 158778-85-9 |
| Molecular Formula | C5 D12 O |
| Quantity | 0.25 g |
| Synonym | 1-Pentan-1,1,2,2,3,3,4,4,5,5,5-d11-ol-d (9 CI), 1-Pentanol-d12, 1-Pentan-d11-ol-d, n-Pentyl-d11 alcohol-OD, n-Pentyl alcohol-d12 |
| InChI Formula | InChI=1 S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2,6 D |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5-undecadeuterio-5-deuteriooxypentane |
| Molecular Weight (g/mol) | 100.2221 |
| Formula Weight | 100.1641 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction