missing translation for 'onlineSavingsMsg'
Learn More

N-p-Benzene Acetonitrile Menthane Carboxamide, TRC

Product Code. 30335876
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335876 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335876 Supplier Toronto Research Chemicals Supplier No. TRCB229870250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-p-Benzene Acetonitrile Menthane Carboxamide
CAS 1187627-98-0
Molecular Formula C19H26N2O
Quantity 250 mg
Synonym (1R,2S,5R)-N-(4(Cyanomethyl)phenyl)menthylcarboxamide, Evercool 180, Evercool G 180, FEMA 4496
InChI Formula InChI=1S/C19H26N2O/c1-13(2)17-9-4-14(3)12-18(17)19(22)21-16-7-5-15(6-8-16)10-11-20/h5-8,13-14,17-18H,4,9-10,12H2,1-3H3,(H,21,22)/t14-,17+,18-/m1/s1
SMILES C(NC1=CC=C(CC#N)C=C1)(=O)[C@H]2[C@H](C(C)C)CC[C@@H](C)C2
IUPAC Name (1R,2S,5R)-N-[4-(cyanomethyl)phenyl]-5-methyl-2-propan-2-ylcyclohexane-1-carboxamide
Molecular Weight (g/mol) 298.42
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.