Learn More
Melford n-Octylglucoside
Product Code. 30018552
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018552 50 g 50g
30018550 25 g 25g
30018549 10 g 10g
30018551 5 g 5g
30018548 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30018552 Supplier Melford Supplier No. N0200750.0
 Restricted Item
Estimated Shipment: 04-11-2026
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Specifications
CAS 29836-26-8
Molecular Formula C14H28O6
Quantity 50 g