Learn More
n-Octyl-d17 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Octanol-d17 |
| CAS | 153336-13-1 |
| Molecular Formula | C8 D17 H O |
| Quantity | 0.5 g |
| Synonym | 1-Octan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17-ol, 1-Octan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-d17-ol (9 CI), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecadeuteriooctan-1-ol, Octyl Alcohol-d17, 1-Hydroxyoctane-d17, Alfol 8-d17, CO 898-d17, CO 898 (solvent)-d17, Caprylic Alcohol-d17, Heptyl Carbinol-d17, Kalcohl 0898-d17, Kalcol 0898-d17, Lorol C 8-98-d17, NSC 9823-d17, Octanol-d17, Octilin-d17, Sipol L8-d17, n-Octan-1-ol-d17, n-Octanol-d17, n-Octyl Alcohol-d17, n-Octyl-d17 alcohol |
| InChI Formula | InChI=1 S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctan-1-ol |
| Molecular Weight (g/mol) | 147.33 |
| Formula Weight | 147.2425 g/mol |
| Show More |