missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octyl-1,1-d2 Alcohol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Octanol-1,1-D2 |
| CAS | 78510-02-8 |
| Molecular Formula | C8 D2 H16 O |
| Quantity | 0.5 g |
| Synonym | 1-Octan-1,1-d2-ol (9 CI, ACI), 1-Octanol-1,1-D2, n-Octyl-1,1-d2 alcohol |
| InChI Formula | InChI=1 S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3/i8D2 |
| SMILES | [2 H]C([2 H])(O)CCCCCCC |
| IUPAC Name | 1,1-dideuteriooctan-1-ol |
| Molecular Weight (g/mol) | 132.24 |
| Formula Weight | 132.148 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction