Learn More
n-Octyl-1,1-d2 Alcohol, CDN
Product Code. 30321506
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321506
Brand: CDN CDND50551G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Octanol-1,1-D2
CAS 78510-02-8
Molecular Formula C8 D2 H16 O
Quantity 1 g
Synonym 1-Octan-1,1-d2-ol (9 CI, ACI), 1-Octanol-1,1-D2, n-Octyl-1,1-d2 alcohol
InChI Formula InChI=1 S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3/i8D2
SMILES [2 H]C([2 H])(O)CCCCCCC
IUPAC Name 1,1-dideuteriooctan-1-ol
Molecular Weight (g/mol) 132.24
Formula Weight 132.148 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity