Learn More
n-Nonyl-d19 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 1-Nonanol-D19 |
| CAS | 349553-86-2 |
| Molecular Formula | C9 D19 H O |
| Quantity | 0.5 g |
| Synonym | 1-Nonan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-d19-ol (9 CI), Nonanol-d19, n-Nonyl-d19 alcohol |
| InChI Formula | InChI=1 S/C9H20O/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecadeuteriononan-1-ol |
| Molecular Weight (g/mol) | 163.3716 |
| Formula Weight | 163.2707 g/mol |
| Show More |