Learn More
N-Nitrosodiisopropylamine- d14, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitrosodiisopropylamine- d14 |
| CAS | 2975202-11-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6D14N2O |
| Quantity | 100 mg |
| Synonym | N-(1-Methylethyl)-N-nitroso-2-propanamine- d14, 1,1'-Dimethyl-N-nitrosodiethylamine- d14, Diisopropyl-N-nitrosamine- d14, Diisopropylnitrosamine- d14, Nitrosodiisopropylamine- d14, NSC 336- d14 |
| InChI Formula | InChI=1S/C6H14N2O/c1-5(2)8(7-9)6(3)4/h5-6H,1-4H3/i1D3,2D3,3D3,4D3,5D,6D |
| SMILES | [2H]C(N(N=O)C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | N,N-bis(propan-2-yl-d7)nitrous amide |
| Molecular Weight (g/mol) | 144.27 |
| Show More |