Learn More
N-Nitroso Tamsulosin Impurity, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitrosotamsulosin |
| CAS | 2892260-31-8 |
| Molecular Formula | C20 H27 N3 O6 S |
| Quantity | 50 mg |
| Synonym | (R)-5-(2-((2-(2-ethoxyphenoxy)ethyl)(nitroso)amino)propyl)-2-methoxybenzenesulfonamide, N-Nitroso Tamsulosin, N-Nitrosotamsulosin |
| InChI Formula | InChI=1S/C20H27N3O6S/c1-4-28-17-7-5-6-8-18(17)29-12-11-23(22-24)15(2)13-16-9-10-19(27-3)20(14-16)30(21,25)26/h5-10,14-15H,4,11-13H2,1-3H3,(H2,21,25,26)/t15-/m1/s1 |
| SMILES | CCOc1ccccc1OCCN(N=O)[C@H](C)Cc2ccc(OC)c(c2)S(=O)(=O)N |
| IUPAC Name | 5-[(2R)-2-[2-(2-ethoxyphenoxy)ethyl-nitrosoamino]propyl]-2-methoxybenzenesulfonamide |
| Molecular Weight (g/mol) | 437.51 |
| Formula Weight | 437.162 |