Learn More
N-Nitroso-L-hydroxyproline, TRC
Product Code. 17932705
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932705 100 mg 100mg
17904125 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17932705 Supplier Toronto Research Chemicals Supplier No. TRCN528600100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30310-80-6
Molecular Formula C5H8N2O4
Molecular Weight (g/mol) 160.13
Synonym (4R)-4-Hydroxy-1-nitroso-L-proline, N-Nitroso-4-hydroxyproline, N-Nitrosohydroxyproline;
SMILES O[C@@H]1C[C@H](N(C1)N=O)C(=O)O
Specifications
InChI Formula InChI=1S/C5H8N2O4/c8-3-1-4(5(9)10)7(2-3)6-11/h3-4,8H,1-2H2,(H,9,10)/t3-,4+/m1/s1
Quantity 100 mg
Formula Weight 160.0484
Shipping Condition Room Temperature
Chemical Name or Material N-Nitroso-L-hydroxyproline