Learn More
N-Nitroso Flecainide, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Flecainide |
| CAS | 2901109-58-6 |
| Molecular Formula | C17H19F6N3O4 |
| Quantity | 10 mg |
| Synonym | N-((1-Nitrosopiperidin-2-yl)methyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide, Benzamide','N-[(1-nitroso-2-piperidinyl)methyl]-2,5-bis(2,2,2-trifluoroethoxy)- (ACI), N-[(1-Nitroso-2-piperidyl)methyl]-2,5-bis(2,2,2-trifluoroethoxy)benzamide |
| InChI Formula | InChI=1S/C17H19F6N3O4/c18-16(19,20)9-29-12-4-5-14(30-10-17(21,22)23)13(7-12)15(27)24-8-11-3-1-2-6-26(11)25-28/h4-5,7,11H,1-3,6,8-10H2,(H,24,27) |
| SMILES | O=C(NCC1CCCCN1N=O)C2=CC(OCC(F)(F)F)=CC=C2OCC(F)(F)F |
| IUPAC Name | N-((1-nitrosopiperidin-2-yl)methyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide |
| Molecular Weight (g/mol) | 443.35 |
| Formula Weight | 443.13 |