Learn More
N-Nitro-S-methylisothiourea, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitro-S-methylisothiourea |
| CAS | 2986-25-6 |
| Molecular Formula | C2H5N3O2S |
| Quantity | 500 mg |
| Synonym | 2-Methyl-1-nitro-2-thiopseudourea, 1-Nitro-2-methylisothiourea, NSC 47544, S-Methyl-1-nitroisothiourea S-Methyl-N-nitroisothiourea S-Methyl-N-nitroisourea, Nitrocarbamimidothioic Acid Methyl Ester, Methyl Nitrocarbamimidothioate |
| InChI Formula | InChI=1S/C2H5N3O2S/c1-8-2(3)4-5(6)7/h1H3,(H2,3,4) |
| SMILES | O=N(NC(SC)=N)=O |
| IUPAC Name | methyl nitrocarbamimidothioate |
| Molecular Weight (g/mol) | 135.14 |
| Formula Weight | 135.01 |