Learn More
N-n-Octyl-D-glucamine, 98%, Thermo Scientific™
Product Code. 15437198
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15437198
Brand: Thermo Scientific Alfa Aesar H60709.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23323-37-7
Molecular Formula C14H31NO5
Molecular Weight (g/mol) 293.404
MDL Number MFCD00134352
InChI Key ZRRNJJURLBXWLL-REWJHTLYSA-N
Synonym n-octyl-d-glucamine, n-octylglucamine, 1-deoxy-1-octylamino-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, n-n-octyl-d-glucamine, 1-octylamino-1-deoxy-d-glucitol, 1-deoxy-1-n-octylamino-d-glucitol, d-glucitol, 1-deoxy-1-octylamino, 1-deoxy-1-octylamine-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol
PubChem CID 90064
IUPAC Name (2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
SMILES CCCCCCCCNCC(C(C(C(CO)O)O)O)O
Specifications
Melting Point 121°C to 124°C
Quantity 250 mg
Optical Rotation −15° (c=1 in Methanol)
Formula Weight 293.4
Percent Purity 98%
Chemical Name or Material N-n-Octyl-D-glucamine