missing translation for 'onlineSavingsMsg'
Learn More

N-n-Octyl-D-glucamine, 98%, Thermo Scientific™

Product Code. 15427198
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427198 1 g 1g
15437198 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427198 Supplier Thermo Scientific Alfa Aesar Supplier No. H60709.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23323-37-7
Molecular Formula C14H31NO5
Molecular Weight (g/mol) 293.404
MDL Number MFCD00134352
InChI Key ZRRNJJURLBXWLL-REWJHTLYSA-N
Synonym n-octyl-d-glucamine, n-octylglucamine, 1-deoxy-1-octylamino-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentaol, n-n-octyl-d-glucamine, 1-octylamino-1-deoxy-d-glucitol, 1-deoxy-1-n-octylamino-d-glucitol, d-glucitol, 1-deoxy-1-octylamino, 1-deoxy-1-octylamine-d-glucitol, 2r,3r,4r,5s-6-octylamino hexane-1,2,3,4,5-pentol
PubChem CID 90064
IUPAC Name (2R,3R,4R,5S)-6-(octylamino)hexane-1,2,3,4,5-pentol
SMILES CCCCCCCCNCC(C(C(C(CO)O)O)O)O

Specifications

Melting Point 121°C to 124°C
Quantity 1 g
Optical Rotation −15° (c=1 in Methanol)
Formula Weight 293.4
Percent Purity 98%
Chemical Name or Material N-n-Octyl-D-glucamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.