Learn More
N,N-Dimethylmethyleneiminium Chloride (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylmethyleneiminium Chloride (Technical Grade) |
| CAS | 30354-18-8 |
| Molecular Formula | C3H8NCl |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C3H8N.ClH/c1-4(2)3;/h1H2,2-3H3;1H/q+1;/p-1 |
| SMILES | [Cl-].C[N+](=C)C |
| IUPAC Name | dimethyl(methylidene)azanium;chloride |
| Molecular Weight (g/mol) | 93.56 |
| Formula Weight | 93.0345 |