missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide-d9, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylacetamide (D9) |
| CAS | 16727-10-9 |
| Molecular Formula | C4D9NO |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C4H9NO/c1-4(6)5(2)3/h1-3H3/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C(=O)C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,2,2-trideuterio-N,N-bis(trideuteriomethyl)acetamide |
| Molecular Weight (g/mol) | 96.176 |
| Formula Weight | 96.1249 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction