missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-d6-tetradecylamine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Tetradecyldimethylamine-D6 (N,N-dimethyl-D6) |
| Molecular Formula | C16 D6 H29 N |
| Quantity | 0.25 g |
| Synonym | N,N-Dimethyl-d6-tetradecylamine, Tetradecyldimethylamine-d6, Tetradecyldimethylamine-D6 (N,N-dimethyl-D6), Tetradecyldi(methyl-d3)amine, Di(methyl-d3)myristylamine, N,N-Di(methyl-d3)-1-tetradecanamine |
| InChI Formula | InChI=1 S/C16H35N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17(2)3/h4-16H2,1-3H3/i2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])N(CCCCCCCCCCCCCC)C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(trideuteriomethyl)tetradecan-1-amine |
| Molecular Weight (g/mol) | 247.493 |
| Formula Weight | 247.315 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction