missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-d6-formamide, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylformamide-D6 (N,N-dimethyl-D6) |
| CAS | 185990-36-7 |
| Molecular Formula | C3 D6 H N O |
| Quantity | 1 g |
| Synonym | Formamide, N,N-di(methyl-d3)- (9 CI), DMF-d6, N,N-Di(methyl-d3)formamide, N,N-Dimethyl-d6-formamide |
| InChI Formula | InChI=1 S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C=O)C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(trideuteriomethyl)formamide |
| Molecular Weight (g/mol) | 79.1308 |
| Formula Weight | 79.0904 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction