Learn More
N,N-Dimethyl-2,2-diphenyl-4-hydroxybutyramide, TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyl-2,2-diphenyl-4-hydroxybutyramide |
| CAS | 37743-13-8 |
| Molecular Formula | C18H21NO2 |
| Quantity | 100 mg |
| Synonym | alpha-(2-Hydroxyethyl)-N,N-dimethyl-alpha-phenylbenzeneacetamide |
| InChI Formula | InChI=1S/C18H21NO2/c1-19(2)17(21)18(13-14-20,15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,20H,13-14H2,1-2H3 |
| SMILES | CN(C)C(=O)C(CCO)(c1ccccc1)c2ccccc2 |
| Molecular Weight (g/mol) | 283.36 |
| Formula Weight | 283.1572 |