Learn More
N,N'-Diethylurea, 97%
Product Code. 11351507
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351507 5 g 5g
11361507 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11351507 Supplier Thermo Scientific Alfa Aesar Supplier No. L02079.06
Request Bulk or Custom Format
CAS: 623-76-7 | C5H12N2O | 116.16 g/mol
N,N'-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 623-76-7
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009028
InChI Key ZWAVGZYKJNOTPX-UHFFFAOYSA-N
Synonym n,n'-diethylurea, sym-diethylurea, urea, n,n'-diethyl, urea, 1,3-diethyl, urea, n,n'-diethyl 9ci, sym-n,n'-diethylurea, n-ethyl ethylamino carboxamide, urea,3-diethyl, urea,n'-diethyl, 1,3-diethyl-urea
PubChem CID 12194
IUPAC Name 1,3-diethylurea
SMILES CCNC(=O)NCC
Specifications
Melting Point 109°C to 113°C
Linear Formula CH3CH2NHCONHCH2CH3
Quantity 5 g
Beilstein 1744741
Solubility Information Soluble in water.
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material N,N'-Diethylurea