Learn More
N,N-Diethylbutyramide, TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Diethylbutyramide |
| CAS | 1114-76-7 |
| Molecular Formula | C8 H17 N O |
| Quantity | 1 g |
| Synonym | N,N-Diethylbutanamide, N,N-Diethyl-n-butyramide, NSC 6005 |
| InChI Formula | InChI=1S/C8H17NO/c1-4-7-8(10)9(5-2)6-3/h4-7H2,1-3H3 |
| SMILES | CCCC(=O)N(CC)CC |
| IUPAC Name | N,N-diethylbutanamide |
| Molecular Weight (g/mol) | 143.227 |
| Formula Weight | 143.131 |