missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(3-aminopropyl-d6)-1,4-butane-d8-diamine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Spermine-d20 |
| Molecular Formula | C10 D20 H6 N4 |
| Quantity | 0.01 g |
| Synonym | N1,N4-Bis(3-aminopropyl-1,1,2,2,3,3-d6)-1,4-butane-1,1,2,2,3,3,4,4-d8-diamine, N,N'-Bis(3-amino-1,1,2,2,3,3-hexadeuteriopropyl)-1,1,2,2,3,3,4,4-octadeuteriobutane-1,4-diamine, N,N'-Bis(3-aminopropyl)-1,4-butanediamine-d20, Spermine-d20, Spermine-D20 |
| InChI Formula | InChI=1 S/C10H26N4/c11-5-3-9-13-7-1-2-8-14-10-4-6-12/h13-14 H,1-12H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])NC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | N,N'-bis(3-amino-1,1,2,2,3,3-hexadeuteriopropyl)-1,1,2,2,3,3,4,4-octadeuteriobutane-1,4-diamine |
| Molecular Weight (g/mol) | 222.464 |
| Formula Weight | 222.341 g/mol |
| Percent Purity | 98 atom % D, min 97% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction