Learn More
N,N’-Bis(2-mercaptobenzoyl)hydrazide, TRC
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis(2-mercaptobenzoyl)hydrazide |
| CAS | 292615-41-9 |
| Molecular Formula | C14H12N2O2S2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C14H12N2O2S2/c17-13(9-5-1-3-7-11(9)19)15-16-14(18)10-6-2-4-8-12(10)20/h1-8,19-20H,(H,15,17)(H,16,18) |
| SMILES | Sc1ccccc1C(=O)NNC(=O)c2ccccc2S |
| IUPAC Name | 2-sulfanyl-N'-(2-sulfanylbenzoyl)benzohydrazide |
| Molecular Weight (g/mol) | 304.39 |