Learn More
N,N-Bis(2-hydroxyethyl)benzenesulfonamide, TRC
Product Code. 30329177
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329177 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30329177 Supplier Toronto Research Chemicals Supplier No. TRCB4438251G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N,N-Bis(2-hydroxyethyl)benzenesulfonamide
CAS 13127-79-2
Molecular Formula C10H15NO4S
Quantity 1 g
Synonym N,N-Bis(2-hydroxyethyl)benzenesulfonamide
InChI Formula InChI=1S/C10H15NO4S/c12-8-6-11(7-9-13)16(14,15)10-4-2-1-3-5-10/h1-5,12-13H,6-9H2
SMILES O=S(C1=CC=CC=C1)(N(CCO)CCO)=O
IUPAC Name N,N-bis(2-hydroxyethyl)benzenesulfonamide
Molecular Weight (g/mol) 245.3
Formula Weight 245.07