missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(2-chloroethyl)phosphorodiamidic Acid Cyclohexylamine Salt, TRC

Product Code. 30335737
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335737 Supplier Toronto Research Chemicals Supplier No. TRCB4189955MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Bis(2-chloroethyl)phosphorodiamidic Acid Cyclohexylamine Salt
CAS 1566-15-0
Molecular Formula C10H24Cl2N3O2P
Quantity 5 mg
Synonym Phosphamide Mustard, NSC 69945
InChI Formula InChI=1S/C6H13N.C4H11Cl2N2O2P/c7-6-4-2-1-3-5-6;5-1-3-8(4-2-6)11(7,9)10/h6H,1-5,7H2;1-4H2,(H3,7,9,10)
SMILES OP(N(CCCl)CCCl)(N)=O.NC1CCCCC1
IUPAC Name amino-[bis(2-chloroethyl)amino]phosphinic acid;cyclohexanamine
Molecular Weight (g/mol) 320.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.