Learn More
N,N'-bis(2,6-Dimethylphenyl)-1,4-piperazinediacetamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2'-(Piperazine-1,4-diyl)bis[N-(2,6-dimethylphenyl)acetamide] |
| CAS | 380204-72-8 |
| Molecular Formula | C24 H32 N4 O2 |
| Quantity | 2.5 g |
| Synonym | 2,2'-(Piperazine-1,4-diyl)bis[N-(2,6-dimethylphenyl)acetamide], N1,N4-bis(2,6-Dimethylphenyl)-1,4-piperazinediacetamide, N,N'-bis(2,6-Dimethylphenyl)-1,4-piperazinediacetamide |
| InChI Formula | InChI=1S/C24H32N4O2/c1-17-7-5-8-18(2)23(17)25-21(29)15-27-11-13-28(14-12-27)16-22(30)26-24-19(3)9-6-10-20(24)4/h5-10H,11-16H2,1-4H3,(H,25,29)(H,26,30) |
| SMILES | Cc1cccc(C)c1NC(=O)CN2CCN(CC(=O)Nc3c(C)cccc3C)CC2 |
| IUPAC Name | 2-[4-[2-(2,6-dimethylanilino)-2-oxoethyl]piperazin-1-yl]-N-(2,6-dimethylphenyl)acetamide |
| Molecular Weight (g/mol) | 408.54 |
| Formula Weight | 408.2525 |