Learn More
N,N’-1,4-Phenylenedi-glycine, TRC
Specifications
Specifications
| Chemical Name or Material | N,N'-1,4-Phenylenedi-glycine |
| CAS | 10097-07-1 |
| Molecular Formula | C10 H12 N2 O4 |
| Quantity | 10 mg |
| Synonym | N,N’-1,4-Phenylenebis-glycine |
| InChI Formula | InChI=1S/C10H12N2O4/c13-9(14)5-11-7-1-2-8(4-3-7)12-6-10(15)16/h1-4,11-12H,5-6H2,(H,13,14)(H,15,16) |
| SMILES | OC(=O)CNc1ccc(NCC(=O)O)cc1 |
| IUPAC Name | 2-[4-(carboxymethylamino)anilino]acetic acid |
| Molecular Weight (g/mol) | 224.21 |
| Formula Weight | 224.0797 |