missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylpiperazine-2,2,3,3,5,5,6,6-d8, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Methylpiperazine-2,2,3,3,5,5,6,6-d8 |
| CAS | 917358-65-7 |
| Molecular Formula | C5 2H8 H4 N2 |
| Quantity | 0.25 g |
| Synonym | 1-Methyl-1,4-piperazine-d8, N-Methylpiperazine-d8, NSC 30195-d8, NSC 30675-d8, Imatinib EP Impurity G-d8 |
| InChI Formula | InChI=1 S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3/i2D2,3D2,4D2,5D2 |
| SMILES | [2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])N(C)C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,5,5,6,6-octadeuterio-1-methylpiperazine |
| Molecular Weight (g/mol) | 108.212 |
| Formula Weight | 108.15 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction