missing translation for 'onlineSavingsMsg'
Learn More

N-Methylglucamine Antimonate, TRC

Product Code. 30467507
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467507 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467507 Supplier Toronto Research Chemicals Supplier No. TRCM31160525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methylglucamine Antimonate
CAS 133-51-7
Molecular Formula C7H17NO5 . HSbO3
Quantity 25 g
Synonym D-Glucitol','1-deoxy-1-(methylamino)-','trioxoantimonate(1-) (9CI), D-Glucitol','1-deoxy-1-(methylamino)-','compd. with antimonic acid (HSbO3) (1:1), Glucitol','1-deoxy-1-(methylamino)-','antimonate(V) (7CI), Glucitol','1-deoxy-1-(methylamino)-','compd. with antimonic acid (HSbO3) (1:1)','D- (8CI), Antimonate (SbO31-)','hydrogen','compd. with 1-deoxy-1-(methylamino)-D-glucitol (1:1) (9CI), Antimonic acid (HSbO3)','compd. with D-1-deoxy-1-(methylamino)glucitol (1:1) (8CI), 2168RP, Glucantim, Glucantime, Meglumine antimonate, Meglumine antimoniate, Methylglucamine antimonate, N-Methylglucamine antimonate, Protostib, RP 2168
InChI Formula InChI=1S/C7H17NO5.H2O.2O.Sb/c1-8-2-4(10)6(12)7(13)5(11)3-9;;;;/h4-13H,2-3H2,1H3;1H2;;;/q;;;;+1/p-1/t4-,5+,6+,7+;;;;/m0..../s1
SMILES CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.O[Sb](=O)=O
IUPAC Name hydroxy(dioxo)-λ^{5}-stibane;(2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 365.98
Formula Weight 365.0071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.