Learn More
N-Methyl-D-alanine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methyl-D-alanine |
| CAS | 29475-64-7 |
| Molecular Formula | C4H9NO2 |
| Quantity | 1 g |
| Synonym | D-Alanine','N-methyl-, Alanine','N-methyl-','D- (8CI), N-Methyl-D-alanine, (2R)-2-(Methylamino)propanoic acid, (R)-2-(Methylamino)propanoic acid, (R)-2-(Methylamino)propionic acid, D-N-Methylalanine |
| InChI Formula | InChI=1S/C4H9NO2/c1-3(5-2)4(6)7/h3,5H,1-2H3,(H,6,7)/t3-/m1/s1 |
| SMILES | CN[C@H](C)C(O)=O |
| IUPAC Name | Methyl-D-alanine |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.06 |