Learn More
N-Isopropylnoradrenochrome, TRC
Specifications
Specifications
| Chemical Name or Material | N-Isopropylnoradrenochrome |
| CAS | 3736-31-0 |
| Molecular Formula | C11H13NO3 |
| Quantity | 100 mg |
| Synonym | 1-Isopropylnoradrenochrome, Aludrinochrome, Isoproterenol Aminochrome, Isoproteroquinone, 2,3-Dihydro-3-hydroxy-1-(1-methylethyl)-1H-indole-5,6-dione |
| InChI Formula | InChI=1S/C11H13NO3/c1-6(2)12-5-11(15)7-3-9(13)10(14)4-8(7)12/h3-4,6,11,15H,5H2,1-2H3 |
| SMILES | CC(C)N1CC(O)C2=CC(=O)C(=O)C=C12 |
| IUPAC Name | 3-hydroxy-1-propan-2-yl-2,3-dihydroindole-5,6-dione |
| Molecular Weight (g/mol) | 207.23 |
| Formula Weight | 207.0895 |