Learn More
n-Hexadecyl-2,2,3,3à16,16,16-d31 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | N-Hexadecanol-D31 |
| CAS | 203633-15-2 |
| Molecular Formula | C16 D31 H3 O |
| Quantity | 0.25 g |
| Synonym | 1-Hexadecan-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d31-ol (9 CI), n-Hexadecanol-D31, n-Hexadecyl-2,2,3,3..0.16,16,16-d31 alcohol, Cetyl alcohol-d31, Cetyl-d31 alcohol, 1-Hexadecanol-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-D31 |
| InChI Formula | InChI=1 S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CO |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-hentriacontadeuteriohexadecan-1-ol |
| Molecular Weight (g/mol) | 273.632 |
| Formula Weight | 273.456 g/mol |
| Show More |