Learn More
n-Hexadecyl-2,2,16,16,16-d5 Alcohol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Hexadecanol-2,2,16,16,16-D5 |
| CAS | 1219799-18-4 |
| Molecular Formula | C16 D5 H29 O |
| Quantity | 0.05 g |
| Synonym | 1-Hexadecan-2,2,16,16,16-d5-ol, [2,2,16,16,16-2H5]Hexadecanol, n-Hexadecanol-2,2,16,16,16-d5, n-Hexadecyl-2,2,16,16,16-d5 alcohol, Cetyl alcohol-β,ω-d5, Cetyl-2,2,16,16,16-d5 alcohol, 1-Hexadecanol-2,2,16,16,16-D5 |
| InChI Formula | InChI=1 S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3/i1D3,15D2 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCCCCCC([2 H])([2 H])CO |
| IUPAC Name | 2,2,16,16,16-pentadeuteriohexadecan-1-ol |
| Molecular Weight (g/mol) | 247.471 |
| Formula Weight | 247.292 g/mol |
| Show More |