Learn More
N-Formylglycine, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2491-15-8 |
| Molecular Formula | C3H5NO3 |
| MDL Number | MFCD00037360 |
| Quantity | 100 g |
| SMILES | O=C(O)CNC=O |
| Molecular Weight (g/mol) | 103.08 |
| Percent Purity | 98% |
| Chemical Name or Material | N-Formylglycine |