Learn More
n-Dodecyl Alcohol-d26, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Dodecanol-D26 |
| CAS | 38086-03-2 |
| Molecular Formula | C12 D26 O |
| Quantity | 0.1 g |
| Synonym | 1-Dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-ol-d (9 CI), 1-Dodecanol-d26, Dodecyl-d25 alcohol-d, 1-Dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-ol (9 CI, ACI), 1-Dodecan-d25-ol-d, 1-Dodecanol-D26, n-Dodecyl-d25 alcohol-O-d |
| InChI Formula | InChI=1 S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13 D |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-pentacosadeuterio-12-deuteriooxydodecane |
| Molecular Weight (g/mol) | 212.494 |
| Formula Weight | 212.362 g/mol |
| Show More |