missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Paroxol Hydrochloride, TRC

Product Code. 17984815
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984815 10 mg 10mg
17990835 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984815 Supplier Toronto Research Chemicals Supplier No. TRCD28218010MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220548-73-2
Molecular Formula C12 H16 F N O . Cl H
Molecular Weight (g/mol) 245.72
Synonym [(3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl]methanol Hydrochloride, (3S,4R)-4-(4-Fluorophenyl)-3-piperidinemethanol hydrochloride, 3-Piperidinemethanol, 4-(4-fluorophenyl)-, hydrochloride (1:1), (3S,4R)-, 3-Piperidinemethanol, 4-(4-fluorophenyl)-, hydrochloride, (3S,4R)- (9CI), (3S,4R)-4-(4-Fluorophenyl)-3-(hydroxymethyl)piperidinium chloride, N-Desmethyl Paroxol hydrochloride, N-Desmethylparoxol hydrochloride
IUPAC Name [(3S,4R)-4-(4-fluorophenyl)piperidin-3-yl]methanol;hydrochloride
SMILES Cl.OC[C@@H]1CNCC[C@H]1c2ccc(F)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H16FNO.ClH/c13-11-3-1-9(2-4-11)12-5-6-14-7-10(12)8-15;/h1-4,10,12,14-15H,5-8H2;1H/t10-,12-;/m0./s1
Formula Weight 245.0983
Chemical Name or Material N-desmethyl Paroxol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.