Learn More
N-Desmethyl Asenapine Hydrochloride, TRC
Product Code. 30357317
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357317 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30357317 Supplier Toronto Research Chemicals Supplier No. TRCD29074010MG
 Restricted Item
Estimated Shipment: 01-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-desmethyl Asenapine hydrochloride
CAS 1170701-78-6
Molecular Formula C16 H14 Cl N O . Cl H
Quantity 10 mg
Synonym 1H-Dibenz[2,3:6,7]oxepino[4,5-c]pyrrole, 5-chloro-2,3,3a,12b-tetrahydro-, hydrochloride (1:1), (3aR,12bR)-rel- (ACI), rel-(3aR,12bR)-5-Chloro-2,3,3a,12b-tetrahydro-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole hydrochloride, N-Desmethylasenapine hydrochloride, trans-5-Chloro-2,3,3a,12b-tetrahydro-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole hydrochloride
InChI Formula InChI=1S/C16H14ClNO.ClH/c17-10-5-6-16-12(7-10)14-9-18-8-13(14)11-3-1-2-4-15(11)19-16;/h1-7,13-14,18H,8-9H2;1H/t13-,14-;/m1./s1
SMILES Cl.Clc1ccc2Oc3ccccc3[C@H]4CNC[C@@H]4c2c1
Molecular Weight (g/mol) 308.2
Formula Weight 307.0531