Learn More
N-Desbutyl-N-propyl Bumetanide, TRC
Product Code. 30366167
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366167 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30366167 Supplier Toronto Research Chemicals Supplier No. TRCD288925100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Desbutyl-N-propyl Bumetanide
CAS 28395-28-0
Molecular Formula C16 H18 N2 O5 S
Quantity 100 mg
Synonym 3-(Aminosulfonyl)-4-phenoxy-5-(propylamino)benzoic Acid, 4-Phenoxy-3-(propylamino)-5-sulfamoylbenzoic Acid, 4-Phenoxy-3-propylamino-5-sulfamoylbenzoic Acid
InChI Formula InChI=1S/C16H18N2O5S/c1-2-8-18-13-9-11(16(19)20)10-14(24(17,21)22)15(13)23-12-6-4-3-5-7-12/h3-7,9-10,18H,2,8H2,1H3,(H,19,20)(H2,17,21,22)
SMILES CCCNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O
IUPAC Name 4-phenoxy-3-(propylamino)-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 350.389
Formula Weight 350.094