Learn More
N-Desbutyl-N-(2-ethylhexyl) Bumetanide (~90% Purity), TRC
Product Code. 30358690
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358690 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30358690 Supplier Toronto Research Chemicals Supplier No. TRCD2890605MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Desbutyl-N-(2-ethylhexyl) Bumetanide
CAS 153012-65-8
Molecular Formula C21 H28 N2 O5 S
Quantity 5 mg
Synonym N-Desbutyl-N-(2-ethylhexyl) Bumetanide (~90% Purity), 3-(Aminosulfonyl)-5-[(2-ethylhexyl)amino]-4-phenoxybenzoic Acid, PF 2825
InChI Formula InChI=1S/C21H28N2O5S/c1-3-5-9-15(4-2)14-23-18-12-16(21(24)25)13-19(29(22,26)27)20(18)28-17-10-7-6-8-11-17/h6-8,10-13,15,23H,3-5,9,14H2,1-2H3,(H,24,25)(H2,22,26,27)
SMILES CCCCC(CC)CNc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O
IUPAC Name 3-(2-ethylhexylamino)-4-phenoxy-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 420.5224
Formula Weight 420.1719