Learn More
N-Des-(4,4-difluorocyclohexanecarboxy)-N-carbobenzyloxy-3-tert-butyldimethylsilyloxymethyl Maraviroc, TRC
Specifications
Specifications
| Chemical Name or Material | N-Des-(4,4-difluorocyclohexanecarboxy)-N-carbobenzyloxy-3-tert-butyldimethylsilyloxymethyl Maraviroc |
| CAS | 1391048-12-6 |
| Molecular Formula | C36H53N5O3Si |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C36H53N5O3Si/c1-26(2)34-39-38-33(25-44-45(6,7)36(3,4)5)41(34)31-22-29-18-19-30(23-31)40(29)21-20-32(28-16-12-9-13-17-28)37-35(42)43-24-27-14-10-8-11-15-27/h8-17,26,29-32H,18-25H2,1-7H3,(H,37,42)/t29?,30?,31?,32-/m0/s1 |
| SMILES | CC(C)c1nnc(CO[Si](C)(C)C(C)(C)C)n1C2CC3CCC(C2)N3CC[C@H](NC(=O)OCc4ccccc4)c5ccccc5 |
| IUPAC Name | benzyl N-[(1S)-3-[3-[3-[[tert-butyl(dimethyl)silyl]oxymethyl]-5-propan-2-yl-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octan-8-yl]-1-phenylpropyl]carbamate |
| Molecular Weight (g/mol) | 631.92 |
| Formula Weight | 631.3918 |