missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Decyl-d21-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Decylamine-D21 |
| CAS | 785003-01-2 |
| Molecular Formula | C10 D21 H2 N |
| Quantity | 0.05 g |
| Synonym | 1-Decan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d21-amine (9 CI), 1-Decan-d21-amine, 1-Decylamine-D21, n-Decyl-d21-amine |
| InChI Formula | InChI=1 S/C10H23N/c1-2-3-4-5-6-7-8-9-10-11/h2-11H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosadeuteriodecan-1-amine |
| Molecular Weight (g/mol) | 178.4257 |
| Formula Weight | 178.3149 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Partager une correction de contenu