Learn More
N-Cbz-D-isoleucine, TRC
Product Code. 30179611
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179611 250 mg 250mg
30179527 50 mg 50mg
30178963 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179611 Supplier Toronto Research Chemicals Supplier No. TRCC227555250MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28862-89-7
Molecular Formula C14 H19 N O4
Molecular Weight (g/mol) 265.3
Synonym N-[(Phenylmethoxy)carbonyl]-D-isoleucine, N-Carboxy-D-isoleucine N-Benzyl Ester, CBZ-D-Isoleucine, Cbz-D-Ile-OH, N-Benzyloxycarbonyl-D-isoleucine, N-Carbobenzoxy-D-isoleucine, N-Carbobenzyloxy-D-isoleucine
IUPAC Name (2R,3R)-3-methyl-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC[C@@H](C)[C@@H](NC(=O)OCc1ccccc1)C(=O)O
Specifications
InChI Formula InChI=1S/C14H19NO4/c1-3-10(2)12(13(16)17)15-14(18)19-9-11-7-5-4-6-8-11/h4-8,10,12H,3,9H2,1-2H3,(H,15,18)(H,16,17)/t10-,12-/m1/s1
Quantity 250 mg
Formula Weight 265.1314
Shipping Condition Room Temperature
Chemical Name or Material N-Cbz-D-isoleucine