missing translation for 'onlineSavingsMsg'
Learn More

N-Caprylic anhydride, TRC

Product Code. 30335921
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335921 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335921 Supplier Toronto Research Chemicals Supplier No. TRCC1757081G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Caprylic anhydride
CAS 623-66-5
Molecular Formula C16 H30 O3
Quantity 1 g
Synonym Octanoic Acid Anhydride
InChI Formula InChI=1S/C16H30O3/c1-3-5-7-9-11-13-15(17)19-16(18)14-12-10-8-6-4-2/h3-14H2,1-2H3
SMILES CCCCCCCC(=O)OC(=O)CCCCCCC
IUPAC Name octanoyl octanoate
Molecular Weight (g/mol) 270.408
Formula Weight 270.219
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.