Learn More
N-Boc-S-ethyl-L-cysteine
Product Code. 11409330
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11409330
Brand: Thermo Scientific Alfa Aesar H50216.06
Request Bulk or Custom Format
CAS: 16947-82-3 | C10H19NO4S | 249.325 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16947-82-3
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.325
MDL Number MFCD00076919
InChI Key IBCCMMVPGKVLAX-ZETCQYMHSA-N
Synonym boc-cys et-oh, boc-s-ethyl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-ethylthio propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-ethylsulfanyl propanoic acid, n-boc-s-ethyl-l-cysteine, s-ethyl-n-t-butoxycarbonyl-l-cysteine, s-ethyl-n-t-butyloxycarbonyl-l-cysteine, n-tert-butoxycarbonyl-s-ethyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-ethyl
PubChem CID 7018740
IUPAC Name (2R)-3-ethylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CCSCC(C(=O)O)NC(=O)OC(C)(C)C
Specifications
Quantity 5 g
Formula Weight 249.33
Chemical Name or Material N-Boc-S-ethyl-L-cysteine